Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF123 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564254
Description
Immunogen sequence: GNMPMLCPPE YMVCFLHRLI SALRYYWDEY KASNPHASFS EEAYIPPQVF YNGKVDYFDL QRLGGLLSHL RKTLKDDLAS KANIVIDPL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Catalytic subunit of the KPC complex that acts as E3 ubiquitin-protein ligase. Required for poly-ubiquitination and proteasome-mediated degradation of CDKN1B during G1 phase of the cell cycle.
Specifications
RNF123 | |
Polyclonal | |
Unconjugated | |
RNF123 | |
BC003945; E3 ubiquitin-protein ligase RNF123; FP1477; Kip1 ubiquitination-promoting complex protein 1; KPC1; RING finger protein 123; RING-type E3 ubiquitin transferase RNF123; RNF123; ubiquitin ligase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63891 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5XPI4 | |
RNF123 | |
Recombinant protein corresponding to Human RNF123. Recombinant protein control fragment (Product #RP-104052). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction