Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF123 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9249R

Description
Catalytic subunit of the KPC complex that acts as E3 ubiquitin-protein ligase. Required for poly-ubiquitination and proteasome-mediated degradation of CDKN1B during G1 phase of the cell cycle.Specifications
RNF123 | |
Polyclonal | |
Unconjugated | |
RNF123 | |
BC003945; E3 ubiquitin-protein ligase RNF123; FP1477; Kip1 ubiquitination-promoting complex protein 1; Kpc1; RING finger protein 123; RING-type E3 ubiquitin transferase RNF123; RNF123; ubiquitin ligase | |
Rabbit | |
Protein A | |
RUO | |
63891 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5XPI4 | |
RNF123 | |
KLH conjugated synthetic peptide derived from human RNF123. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction