Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF126 Mouse anti-Human, Mouse, Rat, Clone: 1E1B10, Proteintech
SDP

Catalog No. 501737065 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7065 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7065 Supplier Proteintech Group Inc Supplier No. 666471IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The specific function of RNF126 is not yet known. The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions.

Specifications

Antigen RNF126
Applications Western Blot
Classification Monoclonal
Clone 1E1B10
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RNF126
Gene Accession No. Q91YL2, Q9BV68
Gene Alias ring finger protein 126, RNF126
Gene Symbols Rnf126
Host Species Mouse
Immunogen RNF126 Fusion Protein Ag13816
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314613, 55658, 70294
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.