Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF126 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112695
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112695 Supplier Invitrogen™ Supplier No. PA5112695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF126 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The specific function of RNF126 is not yet known. The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF126
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RNF126
Gene Accession No. Q91YL2, Q9BV68
Gene Alias 2610010O19Rik; E3 ubiquitin-protein ligase RNF126; ring finger protein 126; RING finger protein 126; E3 ubiquitin-protein ligase RNF126; RNF126
Gene Symbols RNF126
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase RNF126 protein (39-225AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55658, 70294
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.