Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF126 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542321 Supplier Invitrogen™ Supplier No. PA542321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HLFTLPQGYG QFAFGIFDDS FEIPTFPPGA QADDGRDPES RRERDHPSRH Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 100%; Rat: 100%.

The specific function of RNF126 is not yet known. The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF126
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF126
Gene Accession No. Q9BV68
Gene Alias 2610010O19Rik; E3 ubiquitin-protein ligase RNF126; ring finger protein 126; RING finger protein 126; E3 ubiquitin-protein ligase RNF126; RNF126
Gene Symbols RNF126
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RNF126 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55658
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.