Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF128 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523164
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523164 Supplier Invitrogen™ Supplier No. PA523164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The amino acid sequence used as immunogen is 100% homologous in chimpanzee, monkey and orangutan, 94% homologous in cow, 88% homologous in cat and 83% homologous in opossum, rat and mouse.

GRAIL is a type I transmembrane protein that localizes to the endocytic pathway and contains a PA (protease associated) domain and a RING-type zinc finger domain. It is a ubiquitin-protein isopeptide ligase (E3) necessary for the induction of CD4(+) T cell anergy in vivo and induced expression of this protein was observed in anergic CD4 (+) T cells, which suggested a role in the induction of anergic phenotype. It is differentially expressed in naturally occurring and peripherally induced CD25 (+) T regulatory cells and the expression is linked to the conversion of these cells to a regulatory phenotype. Expression of GRAIL in retrovirally transduced T cell hybridomas limits activation-induced IL-2 and IL-4 cytokine production. It also functions in the patterning of the dorsal ectoderm and sensitizes ectoderm to respond to neural-inducing signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF128
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RNF128
Gene Accession No. Q8TEB7, Q9D304
Gene Alias 1300002C13Rik; AI987883; E3 ubiquitin-protein ligase RNF128; Gene related to anergy in lymphocytes protein; goliath-related E3 ubiquitin-protein ligase 1; Grail; Greul1; MNCb-3816; RGD1566282; RING finger protein 128; ring finger protein 128, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF128; RNF128; RP11-150F24.1
Gene Symbols RNF128
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids portion of amino acids 227-269 of human GRAIL.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100054821, 315911, 492051, 66889, 79589
Target Species Human, Mouse, Rat, Canine, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.