Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF128 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107245
Description
Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 93%; Rabbit: 93%; Rat: 92%.
GRAIL is a type I transmembrane protein that localizes to the endocytic pathway and contains a PA (protease associated) domain and a RING-type zinc finger domain. It is a ubiquitin-protein isopeptide ligase (E3) necessary for the induction of CD4(+) T cell anergy in vivo and induced expression of this protein was observed in anergic CD4 (+) T cells, which suggested a role in the induction of anergic phenotype. It is differentially expressed in naturally occurring and peripherally induced CD25 (+) T regulatory cells and the expression is linked to the conversion of these cells to a regulatory phenotype. Expression of GRAIL in retrovirally transduced T cell hybridomas limits activation-induced IL-2 and IL-4 cytokine production. It also functions in the patterning of the dorsal ectoderm and sensitizes ectoderm to respond to neural-inducing signals.
Specifications
| RNF128 | |
| Polyclonal | |
| Unconjugated | |
| RNF128 | |
| 1300002C13Rik; AI987883; E3 ubiquitin-protein ligase RNF128; Gene related to anergy in lymphocytes protein; goliath-related E3 ubiquitin-protein ligase 1; Grail; Greul1; MNCb-3816; RGD1566282; RING finger protein 128; ring finger protein 128, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF128; RNF128; RP11-150F24.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8TEB7 | |
| RNF128 | |
| Synthetic peptide directed towards the C-terminal region of human RNF128. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction