Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568627
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. The specific function of this gene has not yet been determined. Alternatively spliced transcript variants that encode the same protein have been reported. A pseudogene, which is also located on chromosome 3, has been defined for this gene.
Specifications
| RNF13 | |
| Polyclonal | |
| Unconjugated | |
| RNF13 | |
| 2010001H16Rik; E3 ubiquitin-protein ligase RNF13; MGC13689; RING finger protein 13; RING finger protein 13-like protein; RING zinc finger protein; RING-type E3 ubiquitin transferase RNF13; RNF13; RZF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24017 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O54965 | |
| RNF13 | |
| synthetic peptide directed towards the C-terminal region of mouse RNF13. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction