Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598122 Supplier Invitrogen™ Supplier No. PA598122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF13 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. The specific function of this gene has not yet been determined. Alternatively spliced transcript variants that encode the same protein have been reported. A pseudogene, which is also located on chromosome 3, has been defined for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF13
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF13
Gene Accession No. O43567
Gene Alias 2010001H16Rik; E3 ubiquitin-protein ligase RNF13; MGC13689; RING finger protein 13; RING finger protein 13-like protein; RING zinc finger protein; RING-type E3 ubiquitin transferase RNF13; RNF13; RZF
Gene Symbols RNF13
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase RNF13 protein (204-381AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11342
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.