Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF131 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988961
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8961 Supplier Bioss Supplier No. BS9290R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Has E2-dependent E3 ubiquitin-protein ligase activity. Responsible for ubiquitination of cAMP-dependent protein kinase type I and type II-alpha/beta regulatory subunits and for targeting them for proteasomal degradation. Essential for PKA-mediated long-term memory processes.

Specifications

Antigen RNF131
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PJA2
Gene Accession No. O43164
Gene Alias AI447901; AL022700; BOS_8543; E3 ubiquitin-protein ligase Praja2; E3 ubiquitin-protein ligase Praja-2; Kiaa0438; LOC100337733; mKIAA0438; Neurodap1; neurodegeneration associated protein 1; PJA2; praja 2, RING-H2 motif containing; praja 2, RING-H2 motif containing-like; praja ring finger 2; praja ring finger 2, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 2; Praja2; praja-2; protein carrying the RING-H2 sequence motif; RING finger protein 131; RING-type E3 ubiquitin transferase Praja-2; Rnf131
Gene Symbols PJA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PJA2/RNF131.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9867
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.