Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF133 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113294
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113294 Supplier Invitrogen™ Supplier No. PA5113294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF133 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene has no intron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF133
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Rnf133
Gene Accession No. Q8WVZ7
Gene Alias E3 ubiquitin-protein ligase RNF133; Goliath-related E3 ubiquitin-protein ligase 2; Greul2; RING finger protein 133; RING-type E3 ubiquitin transferase RNF133; Rnf133
Gene Symbols Rnf133
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase RNF133 protein (65-167AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168433
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.