Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF135 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598596 Supplier Invitrogen™ Supplier No. PA598596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF135 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by RNF135 contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF135
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF135
Gene Accession No. Q8IUD6
Gene Alias 0610037N03Rik; 2410006N06Rik; E3 ubiquitin-protein ligase RNF135; L13; LRRGT00184; MGC13061; MMFD; REUL; RIG-I E3 ubiquitin ligase; ring finger protein 135; RING finger protein 135; E3 ubiquitin-protein ligase RNF135; RING-type E3 ubiquitin transferase RNF135; Riplet; RNF135; U 2-3-0
Gene Symbols RNF135
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase RNF135 protein (339-432AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84282
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.