Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF139 Polyclonal Antibody
RNF139 Polyclonal Antibody
Supplier: Thermo Scientific PA551534
Description
RNF139 Polyclonal Antibody for Western Blot, IHC

Specifications
RNF139 | |
Polyclonal | |
Unconjugated | |
RNF139 | |
4930555P18Rik; E3 ubiquitin-protein ligase RNF139; HRCA1; MGC31961; multiple membrane spanning receptor TRC8; patched related protein translocated in renal cancer; RCA1; RING finger protein 139; RING finger protein 139; E3 ubiquitin-protein ligase RNF139; RING-type E3 ubiquitin transferase RNF139; RNF139; translocation in renal carcinoma on chromosome 8 protein; TRC8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WU17 | |
RNF139 | |
Recombinant protein corresponding to Human RNF139. Recombinant protein control fragment (Product #RP-96771). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction