Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF139 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518148
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518148 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518148 Supplier Invitrogen™ Supplier No. PA518148
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The protein encoded by this gene is a multi-membrane spanning protein containing a RING-H2 finger. This protein is located in the endoplasmic reticulum, and has been shown to possess ubiquitin ligase activity. This gene was found to be interrupted by a ttranslocation in a family with hereditary renal and non-medulary thyroid cancer. Studies of the Drosophila counterpart suggested that this protein may interact with tumor suppressor protein VHL, as well as with COPS5/JAB1, a protein responsible for the degradation of tumor suppressor CDKN1B/P27KIP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF139
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene RNF139
Gene Accession No. Q8WU17
Gene Alias 4930555P18Rik; E3 ubiquitin-protein ligase RNF139; HRCA1; MGC31961; multiple membrane spanning receptor TRC8; patched related protein translocated in renal cancer; RCA1; RING finger protein 139; RING finger protein 139; E3 ubiquitin-protein ligase RNF139; RING-type E3 ubiquitin transferase RNF139; RNF139; translocation in renal carcinoma on chromosome 8 protein; TRC8
Gene Symbols RNF139
Host Species Goat
Immunogen Synthetic peptide sequence (HTGAAAEEFNDDT) corresponding to the C-terminus amino acids of RNF139 (aa 651-664).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11236
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.