Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF14 Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
SDP

Catalog No. 502543989
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3989 Supplier OriGene Supplier No. CF810955
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNF14 Monoclonal antibody specifically detects RNF14 in Human samples. It is validated for Western Blot

Specifications

Antigen RNF14
Applications Western Blot
Classification Monoclonal
Clone OTI3A4
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Mlx
Gene Accession No. Q9UH92
Gene Alias 2310075C09Rik; 2610005D23Rik; AA986456; androgen receptor associated protein 54; androgen receptor associated protein 54 (androgen receptor coactivator); androgen receptor-associated protein 54; ARA54; AU041447; D18Ertd188e; D7Bwg0165e; E3 ubiquitin-protein ligase RNF14; HFB30; HRIHFB2038; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase RNF14; protein Triad2; RING finger protein 14; RNF14; TRIAD2; Triad2 protein; TRIAD2 type I
Host Species Mouse
Immunogen Full length human recombinant protein of human MLX produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6945
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.