Learn More
Invitrogen™ RNF14 Polyclonal Antibody

Description
Immunogen sequence: GCTMGICSSC NFAFCTLCRL TYHGVSPCKV TAEKLMDLRN EYLQADEANK RLLDQRYGKR VIQKALEEME SKEWLEKNSK SCPCCGTPIE KLDGCNKMTC TGCMQYFCWI CMGSLSRANP YKHFNDPGSP CFNRLFYAVD VDDDIWEDEV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
Specifications
Specifications
| Antigen | RNF14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RNF14 |
| Gene Accession No. | Q9JI90, Q9UBS8 |
| Gene Alias | 2310075C09Rik; 2610005D23Rik; AA986456; androgen receptor associated protein 54; androgen receptor associated protein 54 (androgen receptor coactivator); androgen receptor-associated protein 54; ARA54; AU041447; D18Ertd188e; D7Bwg0165e; E3 ubiquitin-protein ligase RNF14; HFB30; HRIHFB2038; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase RNF14; protein Triad2; RING finger protein 14; RNF14; TRIAD2; Triad2 protein; TRIAD2 type I |
| Gene Symbols | RNF14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.