Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF14 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988921
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8921 Supplier Bioss Supplier No. BS9225R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.

Specifications

Antigen RNF14
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF14
Gene Accession No. Q9UBS8
Gene Alias 2310075C09Rik; 2610005D23Rik; AA986456; androgen receptor associated protein 54; androgen receptor associated protein 54 (androgen receptor coactivator); Androgen receptor-associated protein 54; ARA54; AU041447; BOS_8190; D18Ertd188e; D7Bwg0165e; E3 ubiquitin-protein ligase RNF14; HFB30; HRIHFB2038; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase RNF14; Protein Triad2; RING finger protein 14; Rnf14; Triad2; Triad2 protein; TRIAD2 type I
Gene Symbols RNF14
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF14.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9604
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.