Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF144A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104534
Description
Antibody detects endogenous levels of total RNF144A.
UBCE7IP4 (Ubiquitin-conjugating enzyme 7- interacting protein 4), also known as RNF144A (RING finger protein 144A), KIAA0161 or RNF144, is a 292 amino acid single-pass membrane protein that contains one RING-type zinc finger and two IBR-type zinc fingers. Functioning as an E3 ubiquitin-protein ligase, UBCE7IP4 accepts ubiquitin (in the form of a thioester) from E2 ubiquitin-conjugating enzymes, such as UBC8 and UBCH7, and transfers that ubiquitin residue to target substrates. Via its RING finger, UBCE7IP4 may play a role in protein-DNA and protein-protein interactions throughout the cell.
Specifications
RNF144A | |
Polyclonal | |
Unconjugated | |
RNF144A | |
E3 ubiquitin-protein ligase RNF144A; Kiaa0161; probable E3 ubiquitin-protein ligase RNF144A; RGD1561737; ring finger protein 144; ring finger protein 144A; RNF144; RNF144A; Ubce7ip4; UbcM4-interacting protein 4; ubiquitin conjugating enzyme 7 interacting protein 4; ubiquitin-conjugating enzyme 7-interacting protein 4; Uip4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
108089, 500636, 9781 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P50876, Q925F3 | |
RNF144A | |
A synthesized peptide derived from human RNF144A(Accession P50876), corresponding to amino acid residues H151-M201. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction