Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF144A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110305 Supplier Invitrogen™ Supplier No. PA5110305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELLIKEGLET AISCPDAACP KQGHLQENEI ECMVAAEIMQ RYKKLQFERE VLFDPCRTWC PASTCQAVCQ LQDVGLQTPQ PVQCKACRME FCSTCKASWH PGQGCPETMP ITFLPGETSA AFKMEEDD.

UBCE7IP4 (Ubiquitin-conjugating enzyme 7- interacting protein 4), also known as RNF144A (RING finger protein 144A), KIAA0161 or RNF144, is a 292 amino acid single-pass membrane protein that contains one RING-type zinc finger and two IBR-type zinc fingers. Functioning as an E3 ubiquitin-protein ligase, UBCE7IP4 accepts ubiquitin (in the form of a thioester) from E2 ubiquitin-conjugating enzymes, such as UBC8 and UBCH7, and transfers that ubiquitin residue to target substrates. Via its RING finger, UBCE7IP4 may play a role in protein-DNA and protein-protein interactions throughout the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF144A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RNF144A
Gene Accession No. P50876
Gene Alias E3 ubiquitin-protein ligase RNF144A; Kiaa0161; probable E3 ubiquitin-protein ligase RNF144A; RGD1561737; ring finger protein 144; ring finger protein 144A; RNF144; RNF144A; Ubce7ip4; UbcM4-interacting protein 4; ubiquitin conjugating enzyme 7 interacting protein 4; ubiquitin-conjugating enzyme 7-interacting protein 4; Uip4
Gene Symbols RNF144A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 52-179 of human RNF144A (NP_055561.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500636, 9781
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.