Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF144B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562927 Supplier Invitrogen™ Supplier No. PA562927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGSAGRLHYL AMTAENPTPG DLAPAPLITC KLCLCEQSLD KMTTLQECQC IF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes UBE2L3 and UBE2L6 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates such as LCMT2, thereby promoting their degradation. Induces apoptosis via a p53/TP53-dependent but caspase-independent mechanism. However, its overexpression also produces a decrease of the ubiquitin-dependent stability of BAX, a pro-apoptotic protein, ultimately leading to protection of cell death; But, it is not an anti-apoptotic protein per se.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF144B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF144B
Gene Accession No. Q7Z419
Gene Alias bA528A10.3; BC025007; E130105P19Rik; E3 ubiquitin-protein ligase RNF144B; IBR domain containing 2; IBR domain-containing protein 2; IBRDC2; p53-inducible RING finger protein; p53RFP; PIR2; RING finger protein 144B; Rnf144b
Gene Symbols RNF144B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF144B. Recombinant protein control fragment (Product #RP-103175).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.