Learn More
Invitrogen™ RNF144B Polyclonal Antibody

Description
Immunogen sequence: MGSAGRLHYL AMTAENPTPG DLAPAPLITC KLCLCEQSLD KMTTLQECQC IFCTACLKQY MQLAIREGCG SPITCPDMVC LNHGTLQEAE IACLVPVDQF QLYQRLKFER EVHLDPYRTW CPVADCQTVC PVASSDPGQP VLVECPSCHL KFCSCCKDAW HAEVSCRDSQ PIVLPTEHRA LFGTDAEAPI KQCPVCRVYI; Positive Samples: Mouse kidney, Rat liver, ; Cellular Location: Cytoplasm, Mitochondrion membrane, Single-pass membrane protein.
Specifications
Specifications
| Antigen | RNF144B |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.87 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RNF144B |
| Gene Accession No. | Q8BKD6 |
| Gene Alias | bA528A10.3; BC025007; E130105P19Rik; E3 ubiquitin-protein ligase RNF144B; IBR domain containing 2; IBR domain-containing protein 2; IBRDC2; p53-inducible RING finger protein; p53RFP; PIR2; RING finger protein 144B; Rnf144b |
| Gene Symbols | RNF144B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.