Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF148 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen RNF148
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159516
SDP
View Documents
Novus Biologicals
NBP159516
100 μL
Each of 1 for $480.74
Only null left
Add to Cart
 
Description

Description

RNF148 Polyclonal specifically detects RNF148 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RNF148
Polyclonal
Rabbit
Zinc Finger
MGC35222, ring finger protein 148
RNF148
IgG
34 kDa
Western Blot
Unconjugated
RUO
Q8N7C7
378925
Synthetic peptides corresponding to RNF148(ring finger protein 148) The peptide sequence was selected from the C terminal of RNF148 (NP_932351). Peptide sequence PNSFTRRRSQIKTDVKKAIDQLQLRVLKEGDEELDLNEDNCVVCFDTYKP.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.