Learn More
Invitrogen™ RNF148 Monoclonal Antibody (2D3)

Description
Peptide Sequence: VFPMSHQGTE NIVAVMISNL KGMEILHSIQ KGVYVTVIIE VGRMHMQWVS HYIMYLFTFL AATIAYFYLD CVWRLTPRVP NSFTRRRSQI KTDVKKAIDQ.
Specifications
Specifications
| Antigen | RNF148 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2D3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RNF148 |
| Gene Accession No. | Q8N7C7 |
| Gene Alias | ACVRINP1; AIP1; KIAA0705; MGC35222; RING finger protein 148; ring finger protein 148-like; RN148; Rnf148 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.