Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF148 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542374
Description
Peptide sequence: PNSFTRRRSQ IKTDVKKAID QLQLRVLKEG DEELDLNEDN CVVCFDTYKP Sequence homology: Human: 100%; Rat: 85%; Yeast: 82%.
RNF148 is a single-pass membrane protein. It contains 1 PA (protease associated) domain and 1 RING-type zinc finger. The exact function of RNF148 remains unknown.
Specifications
RNF148 | |
Polyclonal | |
Unconjugated | |
RNF148 | |
ACVRINP1; AIP1; KIAA0705; MGC35222; RING finger protein 148; ring finger protein 148-like; RN148; Rnf148 | |
Rabbit | |
Affinity chromatography | |
RUO | |
378925 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N7C7 | |
RNF148 | |
Synthetic peptide directed towards the C-terminal of human RNF148 (aa 218-267). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction