Learn More
RNF149 Mouse anti-Human, Clone: OTI11H7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810425

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RNF149 is a E3 ubiquitin-protein ligase. Ubiquitinates BRAF, inducing its proteasomal degradation.Specifications
RNF149 | |
Monoclonal | |
Unconjugated | |
RNF149 | |
DNAPTP2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
284996 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI11H7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8NC42 | |
RNF149 | |
Human recombit protein fragment corresponding to amino acids 250-383 of human RNF149 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.