Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF149 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542369
Description
Peptide sequence: LLHTVKHGEK GIDVDAENCA VCIENFKVKD IIRILPCKHI FHRICIDPWL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
RNF149 is a E3 ubiquitin-protein ligase. Ubiquitinates BRAF, inducing its proteasomal degradation.
Specifications
| RNF149 | |
| Polyclonal | |
| Unconjugated | |
| RNF149 | |
| 1600023E10Rik; AV037261; C76763; DNA polymerase-transactivated protein 2; DNAPTP2; E3 ubiquitin-protein ligase RNF149; Gm15832; Goliath-related E3 ubiquitin-protein ligase 4; Greul4; OTTMUSG00000026591; RING finger protein 149; RING-type E3 ubiquitin transferase RNF149; RNF149 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 284996 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NC42 | |
| RNF149 | |
| Synthetic peptide directed towards the C-terminal of human RNF149 (aa 251-300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction