Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF149 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552837 Supplier Invitrogen™ Supplier No. PA552837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLLHTVKHGE KGIDVDAENC AVCIENFKVK DIIRILPCKH IFHRICIDPW LLDHRTCPMC KLDVIKALGY WGEPGDVQEM PAPESPPGRD PAANLSLALP DDDGSDESSP PSASPAESEP QCDPSFKGDA GENT Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%.

RNF149 is a E3 ubiquitin-protein ligase. Ubiquitinates BRAF, inducing its proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF149
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF149
Gene Accession No. Q8NC42
Gene Alias 1600023E10Rik; AV037261; C76763; DNA polymerase-transactivated protein 2; DNAPTP2; E3 ubiquitin-protein ligase RNF149; Gm15832; Goliath-related E3 ubiquitin-protein ligase 4; Greul4; OTTMUSG00000026591; RING finger protein 149; RING-type E3 ubiquitin transferase RNF149; RNF149
Gene Symbols RNF149
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF149. Recombinant protein control fragment (Product #RP-90381).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.