Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF149 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110306 Supplier Invitrogen™ Supplier No. PA5110306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FYYIQRFLYT GSQIGSQSHR KETKKVIGQL LLHTVKHGEK GIDVDAENCA VCIENFKVKD IIRILPCKHI FHRICIDPWL LDHRTCPMCK LDVIKALGYW GEPGDVQEMP APESPPGRDP AANLSLALPD DDGSDDSSPP SASPAESEPQ CDPSFKGDAG ENTALLEAGR SDSRHGGPIS.

RNF149 is a E3 ubiquitin-protein ligase. Ubiquitinates BRAF, inducing its proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF149
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene RNF149
Gene Accession No. Q3U2C5, Q8NC42
Gene Alias 1600023E10Rik; AV037261; C76763; DNA polymerase-transactivated protein 2; DNAPTP2; E3 ubiquitin-protein ligase RNF149; Gm15832; Goliath-related E3 ubiquitin-protein ligase 4; Greul4; OTTMUSG00000026591; RING finger protein 149; RING-type E3 ubiquitin transferase RNF149; RNF149
Gene Symbols RNF149
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 221-400 of human RNF149 (NP_775918.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284996, 67702
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.