Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF150 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558197 Supplier Invitrogen™ Supplier No. PA558197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELHTEKTEC GRYGEHSPKQ DARGEVVMAS SAHDRLACDP NTKFAAPTRG KNWIALIPKG NCTYRDKI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

RNF150 contains a RING-type zinc finger, a motif known to be involved in protein-protein and protein-DNA interactions. The specific function of this protein is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF150
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rnf150
Gene Accession No. Q9ULK6
Gene Alias A630007N06Rik; C030044C12Rik; Greul5; Kiaa1214; LOW QUALITY PROTEIN: RING finger protein 150; mKIAA1214; RGD1304572; RING finger protein 150; Rnf150
Gene Symbols Rnf150
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF150. Recombinant protein control fragment (Product #RP-96734).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57484
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.