Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF151 Monoclonal Antibody (4G8)
GREENER_CHOICE

Catalog No. PIMA524525
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524525 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524525 Supplier Invitrogen™ Supplier No. MA524525
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: HMNKLRKTIG RLEVKCKNAD AGCIVTCPLA HRKGHQDSCP FELTACPNEG CTSQVPRGTL AEHRQHCQQG SQQRCPLGCG ATLDPAERAR HNCYRELHNA WSVRQE.

May be involved in acrosome formation of spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF151
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4G8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RNF151
Gene Accession No. Q2KHN1
Gene Alias 1700010O16Rik; AV047371; LOW QUALITY PROTEIN: RING finger protein 151; ring finger protein 151; RING finger protein 151; LOW QUALITY PROTEIN: RING finger protein 151; Rnf151; Unknown (protein for MGC:133560)
Gene Symbols RNF151
Host Species Mouse
Immunogen RNF151 (NP_056484, 68 a.a. ~ 173 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146310
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.