Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF157 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554544 Supplier Invitrogen™ Supplier No. PA554544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSLSKSTSQN SSVLHEEEDE HSCSESETQL SQRPSVQHLG EECGVTPESE NLTLSSSGAI DQSSCTGTPL SSTISSP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

E3 ubiquitin ligase that ubiquitinates APBB1 for its degradation by the proteasome and thus prevents apoptosis and promotes survival of neurons (PubMed:25342469). Has a dual role in neurons as it is also required for dendrite growth and maintenance for which its ligase activity is not critical (PubMed:25342469). May act as a scaffold molecule to regulate this process (PubMed:25342469). Acts as a downstream effector of the interconnected PI3K and MAPK signaling pathways and thus participates in the regulation of the cell cycle (PubMed:28655764). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF157
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF157
Gene Accession No. Q96PX1
Gene Alias 2610036E23Rik; A130073L17Rik; E3 ubiquitin ligase RNF157; KIAA1917; mKIAA1917; ring finger protein 157; RING-type E3 ubiquitin transferase RNF157; RNF157
Gene Symbols RNF157
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF157. Recombinant protein control fragment (Product #RP-93306).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114804
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.