Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF157 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554529
Description
Immunogen sequence: EEEDGSPTQE GQRTCAFLGM ECDNNNDFDI ASVKALDNKL CSEVCLPGAW QADDNAVSRN AQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
E3 ubiquitin ligase that ubiquitinates APBB1 for its degradation by the proteasome and thus prevents apoptosis and promotes survival of neurons (PubMed:25342469). Has a dual role in neurons as it is also required for dendrite growth and maintenance for which its ligase activity is not critical (PubMed:25342469). May act as a scaffold molecule to regulate this process (PubMed:25342469). Acts as a downstream effector of the interconnected PI3K and MAPK signaling pathways and thus participates in the regulation of the cell cycle (PubMed:28655764). [UniProt].
Specifications
| RNF157 | |
| Polyclonal | |
| Unconjugated | |
| RNF157 | |
| 2610036E23Rik; A130073L17Rik; E3 ubiquitin ligase RNF157; KIAA1917; mKIAA1917; ring finger protein 157; RING-type E3 ubiquitin transferase RNF157; RNF157 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PX1 | |
| RNF157 | |
| Recombinant protein corresponding to Human RNF157. Recombinant protein control fragment (Product #RP-93307). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction