Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF165 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RNF165 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17069620
![]() |
Novus Biologicals
NBP17069620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP170696
![]() |
Novus Biologicals
NBP170696 |
100 μL |
Each for $487.50
|
|
|||||
Description
RNF165 Polyclonal specifically detects RNF165 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RNF165 | |
Polyclonal | |
Rabbit | |
ARKL2, ring finger protein 165 | |
RNF165 | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
494470 | |
Synthetic peptides corresponding to RNF165(ring finger protein 165) The peptide sequence was selected from the middle region of RNF165. Peptide sequence SSTQMVVHEIRNYPYPQLHFLALQGLNPSRHTSAVRESYEELLQLEDRLG. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title