Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF165 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559649 Supplier Invitrogen™ Supplier No. PA559649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALHQQYLLQQ QLLEAQHRRL VSHPRRSQER VSVHPHRLHP SFDFGQLQTP QPRYLAEGTD WDLSVDAGLS PAQFQVRPIP QHYQHYLATP RMHHFPRNSS STQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Encoded in regions involved in pericentric inversions in patients with bipolar affective disorder. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF165
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF165
Gene Accession No. Q6ZSG1
Gene Alias 2900024M11Rik; AI427432; Akd2; Ark2c; arkadia 2; Arkadia-like protein 2C; ARKL2; E3 ubiquitin-protein ligase RNF165; G630064H08Rik; Gm96; RGD1560744; RING finger protein 165; RNF111L2; RNF165
Gene Symbols RNF165
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF165. Recombinant protein control fragment (Product #RP-98242).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494470
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.