Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF166 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576363 Supplier Invitrogen™ Supplier No. PA576363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RNF166 (RING finger protein 166) is a 237 amino acid protein that contains one RING-type zinc finger and a UIM (ubiquitin-interacting motif) repeat. Existing as two alternatively spliced isoforms, RNF166 is encoded by a gene that maps to human chromosome 16q24.3. Chromosome 16 encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF166
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF166
Gene Accession No. Q3U9F6, Q6J1I7, Q96A37
Gene Alias 1110031E24Rik; AW555115; E3 ubiquitin-protein ligase RNF166; RING finger protein 166; RING-type E3 ubiquitin transferase RNF166; RNF166; Zfp313l; zinc finger protein 313; zinc finger protein 228
Gene Symbols RNF166
Host Species Rabbit
Immunogen Synthetic peptide of human RNF166.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115992, 365022, 68718
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.