Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF166 Polyclonal Antibody

RNF166 Polyclonal Antibody
Supplier: Thermo Scientific PA5111169
Description
RNF166 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| RNF166 | |
| Polyclonal | |
| Unconjugated | |
| RNF166 | |
| 1110031E24Rik; AW555115; E3 ubiquitin-protein ligase RNF166; RING finger protein 166; RING-type E3 ubiquitin transferase RNF166; RNF166; Zfp313l; zinc finger protein 313; zinc finger protein 228 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115992 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.40 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96A37 | |
| RNF166 | |
| Recombinant Protein corresonding to Human RNF166. Recombinant protein control fragment (Product #RP-98678). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction