Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF167 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103821 Supplier Invitrogen™ Supplier No. PA5103821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RNF167.

May act as an E3 ubiquitin-protein ligase, or as part of the E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, such as UBE2E1, and then transfers it to substrates, such as SLC22A18. May play a role in growth regulation involved in G1/S transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF167
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RNF167
Gene Accession No. Q5XIL0, Q91XF4, Q9H6Y7
Gene Alias 0610010G05Rik; 5730408C10Rik; AU019305; AV328608; E3 ubiquitin-protein ligase RNF167; LP2254; novel PA domain and C3HC4 type zinc finger (RING finger) containing protein; RGD1305972; RING finger protein 167; RING105; RING-type E3 ubiquitin transferase RNF167; Rnf167
Gene Symbols RNF167
Host Species Rabbit
Immunogen A synthesized peptide derived from human RNF167(Accession Q9H6Y7), corresponding to amino acid residues K266-T316.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26001, 360554, 70510
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.