Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF167 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988925
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8925 Supplier Bioss Supplier No. BS9232R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May act as an E3 ubiquitin-protein ligase, or as part of the E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, such as UBE2E1, and then transfers it to substrates, such as SLC22A18. May play a role in growth regulation involved in G1/S transition.

Specifications

Antigen RNF167
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF167
Gene Accession No. Q9H6Y7
Gene Alias 0610010G05Rik; 5730408C10Rik; AU019305; AV328608; E3 ubiquitin-protein ligase RNF167; LP2254; novel PA domain and C3HC4 type zinc finger (RING finger) containing protein; RGD1305972; RING finger protein 167; RING105; RING-type E3 ubiquitin transferase RNF167; RNF167
Gene Symbols RNF167
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF167.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26001
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.