Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF168 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | RNF168 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RNF168 Polyclonal specifically detects RNF168 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RNF168 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
E3 ubiquitin-protein ligase RNF168, EC 6.3.2, EC 6.3.2.-, FLJ39749, ring finger protein 168FLJ35794 | |
RNF168 | |
IgG | |
Affinity Purified |
Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
165918 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LTPKSQFGSASHSEAVQEVRKDSVSKDIDSSDRKSPTGQDTEIEDMPTLSPQISLGVGEQGADSSIESPMPWLCACGAEWYHEGNVKTRPSNHGK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title