Learn More
Invitrogen™ RNF168 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20738 can be used with blocking peptide PEP-0852.
Specifications
Specifications
| Antigen | RNF168 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RNF168 |
| Gene Accession No. | Q80XJ2, Q8IYW5 |
| Gene Alias | 3110001H15Rik; E3 ubiquitin-protein ligase RNF168; FLJ35794; hRNF168; RING finger protein 168; ring finger protein 168, E3 ubiquitin protein ligase; ring fnger protein 168; RING-type E3 ubiquitin transferase RNF168; RNF168 |
| Gene Symbols | RNF168 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.