Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF169 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566164
Description
Immunogen sequence: KAKVTTMTPAS NPIIGVLLST QNNRCVSAPD LTIEKRLPFS SLSSLASLHK PERSVSPESN DSISEELNHF KPIVCSPCTP PKRLPDGRVL SPLIIKSTPR NLN Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Probable E3 ubiquitin-protein ligase that acts as a negative regulator of double-strand breaks (DSBs) repair following DNA damage. Recruited to DSB repair sites by recognizing and binding ubiquitin catalyzed by RNF168 and competes with TP53BP1 and BRCA1 for association with RNF168-modified chromatin, thereby acting as a negative regulator of DSBs repair. E3 ubiquitin-protein ligase activity is not required for regulation of DSBs repair.
Specifications
RNF169 | |
Polyclonal | |
Unconjugated | |
RNF169 | |
2900057K09Rik; E3 ubiquitin-protein ligase RNF169; KIAA1991; RING finger protein 169; RING finger protein 169; E3 ubiquitin-protein ligase RNF169; RING-type E3 ubiquitin transferase RNF169; RNF169 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
254225 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCN4 | |
RNF169 | |
Recombinant Human RNF169. Recombinant protein control fragment (Product #RP-106847). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction