Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF170 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988949
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8949 Supplier Bioss Supplier No. BS9260R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF170 is a multi-pass membrane protein. It contains 1 RING-type zinc finger. The exact function of RNF170 remains unknown.

Specifications

Antigen RNF170
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF170
Gene Accession No. Q96K19
Gene Alias 6720407G21Rik; ADSA; AI481227; BOS_23912; E3 ubiquitin-protein ligase RNF170; LOC100347695; Putative LAG1-interacting protein; RING finger protein 170; RING-type E3 ubiquitin transferase RNF170; RNF170; RNF170 protein-like; SNAX1
Gene Symbols RNF170
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF170.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81790
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.