Learn More
Invitrogen™ RNF175 Monoclonal Antibody (4E9)

Description
Peptide Sequence: FYSVSRLPTR SLSDNICAVC GQKIIVELDE EGLIENTYQL SCNHVFHEFC IRGWCIVGKK QTCPYCKEKV DLKRMISNPW ERTHFLYGQI L.
Specifications
Specifications
| Antigen | RNF175 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4E9 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RNF175 |
| Gene Accession No. | Q8N4F7 |
| Gene Alias | RING finger protein 175; RNF175 |
| Gene Symbols | RNF175 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.