Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF180 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305783AF700
Description
RNF180 Polyclonal specifically detects RNF180 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RNF180 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| E3 ubiquitin-protein ligase RNF180, EC 6.3.2.-, MGC120328, ring finger protein 180MGC120326 | |
| Produced in rabbits immunized with E. coli-derived Human RNF180 fragment. | |
| 0.1 mL | |
| Zinc Finger | |
| 285671 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction