Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF181 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561153
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561153 Supplier Invitrogen™ Supplier No. PA561153
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASYFDEHDCE PSDPEQETRT NMLLELARSL FNRMDFEDLG LVVDWDHHLP PPAAKTVVEN LPRTVIRGSQ AELKCP Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.

RNF181 binds the integrin alpha-IIb (ITGA2B)/beta-3 (ITGB3) complex and has E3 ubiquitin ligase activity (Brophy et al., 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF181
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rnf181
Gene Accession No. Q9P0P0
Gene Alias 1700047F16Rik; 2500002L14Rik; 6230415N01Rik; C77350; E3 ubiquitin-protein ligase RNF181; HSPC238; RING finger protein 181; RING-type E3 ubiquitin transferase RNF181; RNF181
Gene Symbols Rnf181
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF181. Recombinant protein control fragment (Product #RP-96192).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.