Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF182 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552925 Supplier Invitrogen™ Supplier No. PA552925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLECCHRVCA KCLYKIIDFG DSPQGVIVCP FCRFETCLPD DEVSSLPDDN NILVNLTCGG KGKKCLPENP TELLLTPKRL ASLVSPSHTS SNCLVITIME VQRESSPSLS STPVVEFYRP ASFDSVTTVS HNWTVWNCTS LL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

E3 ubiquitin-protein ligase that mediates the ubiquitination of ATP6V0C and targets it to degradation via the ubiquitin-proteasome pathway (PubMed:18298843). Plays also a role in the inhibition of TLR-triggered innate immune response by mediating 'Lys'-48-linked ubiquitination and subsequent degradation of NF-kappa-B component RELA (PubMed:31432514). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF182
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF182
Gene Accession No. Q8N6D2
Gene Alias E3 ubiquitin-protein ligase RNF182; ring finger protein 182; RING-type E3 ubiquitin transferase RNF182; RNF182
Gene Symbols RNF182
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF182. Recombinant protein control fragment (Product #RP-88711).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221687
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.