Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF183 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563466 Supplier Invitrogen™ Supplier No. PA563466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELEAECPVCW NPFNNTFHTP KMLDCCHSFC VECLAHLSLV TPARRRLLCP LCRQPTVLAS GQPVTDLPTD Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.

Acts as a E3 ubiquitin ligase catalyzing the covalent attachment of ubiquitin moieties onto substrate proteins (PubMed:29507230). Triggers apoptosis in response to prolonged ER stress by mediating the polyubiquitination and subsequent proteasomal degradation of BCL2L1 (PubMed:29507230). May collaborate with FATE1 to restrain BIK protein levels thus regulating apoptotic signaling (PubMed:26567849). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF183
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF183
Gene Accession No. Q96D59
Gene Alias 5830442J12Rik; E3 ubiquitin-protein ligase RNF183; ring finger protein 183; Rnf183
Gene Symbols RNF183
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF183. Recombinant protein control fragment (Product #RP-99866).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138065
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.