Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF185 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107016 Supplier Invitrogen™ Supplier No. PA5107016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 100%.

RNF185 is a E3 ubiquitin-protein ligase that regulates selective mitochondrial autophagy by mediating Lys-63-linked polyubiquitination of BNIP1. RNF185 acts in the endoplasmic reticulum (ER)-associated degradation (ERAD) pathway, which targets misfolded proteins that accumulate in the endoplasmic reticulum (ER) for ubiquitination and subsequent proteasome-mediated degradation. RNF185 protects cells from ER stress-induced apoptosis. is responsible for the cotranslational ubiquitination and degradation of CFTR in the ERAD pathway, and is preferentially associates with the E2 enzymes UBE2J1 and UBE2J2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF185
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene RNF185
Gene Accession No. Q96GF1
Gene Alias BSK65-MONO1; BSK65-MONO2; BSK65-PANC1; BSK65-PANC2; BSK65-TEST1; BSK65-TEST2; BSK65-TEST3; E3 ubiquitin-protein ligase RNF185; FLJ38628; I79_017427; ring finger protein 185; RING-type E3 ubiquitin transferase RNF185; rn185; Rnf185
Gene Symbols RNF185
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RNF185 (aa 74-124).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91445
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.