Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF185 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988950
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8950 Supplier Bioss Supplier No. BS9261R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF185 is a E3 ubiquitin-protein ligase that regulates selective mitochondrial autophagy by mediating Lys-63-linked polyubiquitination of BNIP1. RNF185 acts in the endoplasmic reticulum (ER)-associated degradation (ERAD) pathway, which targets misfolded proteins that accumulate in the endoplasmic reticulum (ER) for ubiquitination and subsequent proteasome-mediated degradation. RNF185 protects cells from ER stress-induced apoptosis. is responsible for the cotranslational ubiquitination and degradation of CFTR in the ERAD pathway, and is preferentially associates with the E2 enzymes UBE2J1 and UBE2J2.

Specifications

Antigen RNF185
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF185
Gene Accession No. Q96GF1
Gene Alias BSK65-MONO1; BSK65-MONO2; BSK65-PANC1; BSK65-PANC2; BSK65-TEST1; BSK65-TEST2; BSK65-TEST3; E3 ubiquitin-protein ligase RNF185; FLJ38628; RING finger protein 185; RING-type E3 ubiquitin transferase RNF185; RNF185
Gene Symbols RNF185
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF185.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91445
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.