Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF187 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556340 Supplier Invitrogen™ Supplier No. PA556340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVMDRRKKAL TDYKKLRAFF VEEEEHFLQE AEKEEGLPED ELADPTERFR SLLQAVSELE KKHRNLGLSM LLQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

E3 ubiquitin-protein ligase that acts as a coactivator of JUN-mediated gene activation in response to growth factor signaling via the MAP3K1 pathway, independently from MAPK8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF187
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF187
Gene Accession No. Q5TA31
Gene Alias 2410016F01Rik; AB030190; E3 ubiquitin-protein ligase RNF187; protein RNF187; RACO1; RACO-1; RGD1308636; RING domain AP1 coactivator 1; ring finger protein 187; RING-type E3 ubiquitin transferase RNF187; Rnf187
Gene Symbols RNF187
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF187. Recombinant protein control fragment (Product #RP-94443).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.